missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Axygen™ Single Well High Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
$848.22 - $1100.84
Specifications
Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
---|---|
Color | Clear |
Material | Polypropylene |
Type | Reagent Reservoir |
Disposable | Single-use |
Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Channels | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Channels | Price | Quantity | |||
14-222-418
|
Axygen™
RESSW384HPSI |
282 mL | Sterile | 384 |
Case of 25 Each for $967.88
|
|
|||
14-222-422
|
Axygen™
RESSW8HPSI |
290 mL | Sterile | 8 |
Case of 25 Each for $967.88
|
|
|||
14-222-412
|
Axygen™
RESSW12HPSI |
290 mL | Sterile | 12 |
Case of 25 Each for $1,100.84
|
|
|||
50-400-726
|
Axygen™
RESSW8HP |
290 mL | Non-sterile | 8 |
Case of 25 Each for $1,001.24
|
|
|||
50-400-720
|
Axygen™
RESSW12HP |
290 mL | Non-sterile | 12 |
Case of 25 Each for $848.22
|
|
|||
14-222-424
|
Axygen™
RESSW96HPSI |
240 mL | Sterile | 96 |
Case of 25 Each for $1,000.56
|
|
|||
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
Polypropylene | |
Single-use |
Clear | |
Reagent Reservoir |