missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

β-Sitosterol, Practical grade, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210288680

 View more versions of this product

Catalog No. ICN10288680


Explore available promotions

Description

Description

Specifications

Specifications

β-Sitosterol
White
100 g
KZJWDPNRJALLNS-VJSFXXLFSA-N
(3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
222284
414.7
Powder
83-46-5
C29H50O
beta-sitosterol, sitosterol, cupreol, quebrachol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C
414.718
CHEBI:27693
Practical

Chemical Identifiers

83-46-5
414.718
beta-sitosterol, sitosterol, cupreol, quebrachol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
CHEBI:27693
CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C
C29H50O
KZJWDPNRJALLNS-VJSFXXLFSA-N
222284
(3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Videos
SDS
Documents

Documents

Promotions

Promotions