missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Laxco™ SLi3 PRO Series Inverted Compound Microscope

The SLi3 PRO Series Inverted Compound Microscope is an upgradable trinocular microscope. Phase-contrast, fluorescent, and phase-contrast fluorescent options are available with complementing filters included.

$17308.65 - $43878.16

Specifications

Condenser Type NA 0.3 (WD 72 mm)
Nosepiece Quintuple Nosepiece
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Stage Fixed
Eyepiece WF 10X/22 mm
View More Specs

Products 3
Catalog Number Mfr. No. Model Product Type Objective Types Price Quantity  
Catalog Number Mfr. No. Model Product Type Objective Types Price Quantity  
SLI3PFL1
View Documents
Laxco™
SLI3PFL1
SLi3P-FL1 Inverted Fluorescent Microscope Infinity LWD Plan Fluorescent Objective (4X, 10X, 40X) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SLI3PPH1
View Documents Promotion Details
Laxco™
SLI3PPH1
SLi3P-PH1 Inverted Phase-contrast Microscope Infinity LWD Plan Achromatic Objective (4X) and Positive Phase Contrast (10X, 20X) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SLI3PFLP
View Documents
Laxco™
SLI3PFLP
SLi3P-FLP Inverted Phase-contrast Fluorescent Microscope Infinity LWD Plan Fluorescent Objective (4X, 10X) and Infinity LWD Plan Positive Phase Fluorescent Objective (20X, 40X) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Infinity corrected optical system
  • Trinocular 45° inclined head rotates 360°
  • Adjustable interpupillary distance of 50 mm to 75 mm with 100:100 image split
  • High eye-point, wide-field plan eyepiece (10x/22 mm FOV) with adjustable eyecups
  • Coarse and fine low-position coaxial focusing mechanism with tension adjustment and upper limit
  • N.A. 0.3 condenser with 72 mm working distance which can be removed to view specimens in large vessel
  • Quintuple nosepiece
  • Upgrade kits available (sold separately)
Specifications

Specifications

NA 0.3 (WD 72 mm)
5 years optical and mechanical, 1 year electrical from date of purchase
WF 10X/22 mm
Trinocular
High
Quintuple Nosepiece
Fixed
LED
SLi3 PRO Series
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'