missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Devices, 2K MWCO
Perform sample buffer exchange with these 2000-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.
Supplier: Thermo Scientific™ 69553
Description
Slide-A-Lyzer MINI Dialysis Devices, 2K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules larger than 2 kDa in volumes of 10 to 100 μL. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices.
Features of Slide-A-Lyzer MINI Dialysis Devices, 2K MWCO:• Flexible format—enables user to dialyze a single sample (tube) or multiple samples (25 array)
• Excellent sample recovery—low-binding plastic and small membrane surface area minimize sample loss compared to filtration and resin systems
• One-step protocol—does not require laborious procedures or expensive equipment
• 100% leak-tested—innovative design does not permit 'wicking' that can occur in homemade devices
• Time saving—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes
Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. They are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.
The 0.1-mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL microcentrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker. Using the 0.1-mL MINI device, 100 μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes.
As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.
Specifications
Cup, Microcentrifuge | |
2.0 kDa | |
MINI Dialysis Device | |
Buffer Exchange, Protein | |
250 Devices |
Slide-A-Lyzer™ | |
Cellulose Membrane, Dialysis | |
MINI Dialysis Device, 0.1 mL | |
0.1 mL | |
Store at room temperature |