missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Devices, 3.5K MWCO

Perform sample buffer exchange with these 3500-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.

$197.70 - $1,251.30

Specifications

Product Line Slide-A-Lyzer™
MWCO 3.5 kDa
Membrane Cellulose Membrane, Dialysis
Format MINI Dialysis Device
Purification Target Buffer Exchange, Protein
View More Specs

Products 4
Catalog Number Mfr. No. Product Type Sample Volume (Metric) Sufficient For Price Quantity  
Catalog Number Mfr. No. Product Type Sample Volume (Metric) Sufficient For Price Quantity  
PI69550
View Documents
Thermo Scientific™
69550
Cup, Microcentrifuge 0.1 mL 50 Sample N/A
 
PI69552
View Documents
Thermo Scientific™
69552
Cup, Microcentrifuge 0.1 mL 250 Samples N/A
 
PI69558
View Documents
Thermo Scientific™
69558
Cup, Microcentrifuge Kit 0.1 mL 10 Samples N/A
 
PI88403
View Documents
Thermo Scientific™
88403
Cup, Conical 2 mL 25 Samples N/A
 
Description

Description

Slide-A-Lyzer MINI Dialysis Devices, 3.5K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules >3.5kDa in volumes from 10 μL to 2 mL. They were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices.

Slide-A-Lyzer MINI Dialysis Devices are manufactured in a clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use. These 3.5K MWCO dialysis devices are available in three sizes (0.1, 0.5, and 2 mL capacities), and are designed to allow easy sample addition and removal using a standard laboratory pipette. The MINI devices are effective and convenient for accomplishing typical sample desalting or buffer exchange in 4–8 hours. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters, and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with common chemicals and buffers.

Features of Slide-A-Lyzer MINI Dialysis Devices, 3.5K MWCO:
Three convenient sizes—dialysis devices for 0.1, 0.5, and 2 mL samples fit standard 1.5-mL micro centrifuge, 15- and 50-mL conical tubes, respectively
Excellent sample recovery—low-binding plastic and small membrane surface area minimize sample loss
One-step protocol—pipette sample into the MINI device and place in tube containing the dialysis buffer; no laborious assembly, device preparation, or expensive equipment are required
100% leak-tested—innovative design does not permit ‘wicking’ that can occur in home-made devices
Time saving—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes, which improves sample resolution, quantification, and conjugation

The 0.5- and 2-mL MINI dialysis devices fit in and are capped by inserting them into the included 15- and 50-mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI dialysis devices, low molecular weight contaminant removal and buffer exchange and desalting can be accomplished within 4–8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.

The 0.1 mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL micro-centrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker.

Using the 0.1 mL MINI Device, 100 μL of pH 2.8 glycine buffer can be equilibrated to 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes. As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.

Specifications

Specifications

Slide-A-Lyzer™
Cellulose Membrane, Dialysis
Buffer Exchange, Protein
3.5 kDa
MINI Dialysis Device
Store at room temperature
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'