missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Slide-A-Lyzer™ Accessory Float Buoys and Syringes
Slide-A-Lyzer Float Buoys and Syringes are available to support a variety of dialysis cassettes for volumes of 0.1 to 20 mL.
$76.75 - $92.45
Specifications
Content And Storage | Store at room temperature. |
---|---|
Membrane | Dialysis |
Format | Cassette, Accessory |
Product Line | Slide-A-Lyzer™ |
Catalog Number | Mfr. No. | Capacity (Metric) | Product Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Product Type | Price | Quantity | ||||
PI66430
|
Thermo Scientific™
66430 |
For 0.5 to 3 mL Cassettes | Cassette Float Buoy |
Pack of 10 Each for $92.45
|
|
||||
PI66432
|
Thermo Scientific™
66432 |
For 12 mL Cassettes | Cassette Float Buoy |
Pack of 8 Each for $83.89
|
|
||||
PI66490
|
Thermo Scientific™
66490 |
5 mL | Cassette Syringe |
Pack of 10 Each for $76.75
|
|
||||
PI66493
|
Thermo Scientific™
66493 |
20 mL | Cassette Syringe |
Pack of 10 Each for $79.68
|
|
||||
PI66494
|
Thermo Scientific™
66494 |
1 mL | Cassette Syringe |
Pack for $82.27
|
|
||||
Description
Slide-A-Lyzer Dialysis Cassette Syringes are designed for efficient loading of samples into Slide-A-Lyzer cassettes and removal of sample after dialysis. Cassette Float Buoy accessories are for use with Slide-A-Lyzer Dialysis Cassettes used to remove low molecular weight contaminants and perform buffer exchange, desalting, and concentration.
Minimize sample loss into and out of Slide-A-Lyzer Dialysis Cassettes using Slide-A-Lyzer Dialysis Cassette Syringes for 1-mL to 20-mL sample volumes. Slide-A-Lyzer Cassette Float Buoys are designed for floating 0.5- to 12-mL cassettes in dialysate.Features of Slide-A-Lyzer Cassette Accessories include:
• Syringes are appropriately sized for different sample volumes
• Buoys keep stir bar from tearing dialysis membrane
• Matched to Slide-A-Lyzer Cassette for a secure fit and minimization of sample loss
• Chemically inert
Specifications
Store at room temperature. | |
Dialysis | |
Cassette, Accessory | |
Slide-A-Lyzer™ |