missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 3.5K MWCO

Remove salts and contaminants, and maximize sample recovery using this semi-permeable membrane with a 3,500-Dalton protein MW cut-off (MWCO) cassette that is easier to use than traditional dialysis tubing.

Supplier:  Thermo Scientific™ 66335

 View more versions of this product

Catalog No. PI66335


Explore available promotions

Description

Description

Slide-A-Lyzer Dialysis Cassettes (3.5K MWCO) facilitate simple and effective removal of salts and small contaminants from proteins and other macromolecules larger than 3,500 Daltons. They can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.

Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample loss during the dialysis procedure. Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume from any one of the four corner injection ports
Four cassette sizes—select the cassette that best suits your sample volume (0.5 to 30 mL)
Color-coded—pink cassette frame specifies cassettes with the 3,500 MW pore size membrane

Membrane specifications:
Composition: regenerated cellulose synthesized by the viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: none
Sulfur content: 0.1–0.15%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL

Specifications

Specifications

Store at room temperature
Cellulose Membrane, Dialysis
3.5 kDa
1 Kit (10 Cassettes)
Buffer Exchange, Protein
Cassette
Slide-A-Lyzer™
0.5 mL
Videos
SDS
Documents

Documents

Promotions

Promotions