Learn More
Epredia™ Slide Baskets and Hangers for Epredia™ ClearVue™ Coverslipper
Use these slide baskets and hangers with the Epredia™ ClearVue™ Coverslipper to make it compatible with any automated slide stainer.
$516.88 - $757.67
Specifications
| For Use With (Equipment) | Epredia ClearVue Coverslipper |
|---|---|
| Quantity | 5 Pk. |
| Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
|
28-618-375
|
Epredia™
A79210064 |
Gemini Basket, Black Slider |
Each for $695.44
|
|
|||||
|
28-618-376
|
Epredia™
A79210065 |
Gemini Basket, White Slider |
Each for $757.67
|
|
|||||
|
28-618-377
|
Epredia™
A79210066 |
Leica Basket, Black Slider |
Each for $631.59
|
|
|||||
|
A79210072
|
Epredia™
A79210072 |
Leica Basket, Black Slider |
Pack of 5 Each for $516.88
|
|
|||||
Description
- Allow equipment compatibility with a variety of different stainers
- Can be loaded in mix-or-match fashion
- Sold in sets of five
Specifications
| Epredia ClearVue Coverslipper | |
| 5 Pk. |