Dynalon Slide Storing System, 4 Drawer
Semi-transparent ice color gives an attractive appearance while the metal slide storing cabinet structure makes it stable.
Supplier: Dynalon 209464
Description
- Slide Storage System is designed for the classification and storage of microscope slides.
- It is composed of 4 sliding drawers, a closing cover and four fixed support feet.
- Four swivel castors to guarantee mobility are available as an accessory.
- This is a completely new storing system, made of impact-resistant PMMA and steel.
- Transparent cabinet for slide storage is real design object that will also be appreciated by users who are attentive to the aesthetic aspect.
- The semi-transparent ice color gives an attractive appearance while the metal structure makes it stable, while ensuring the value of lightness, functionality, strength and durability.
- Each single drawer can hold up to 13 supports for slides and transparent cover plates.
- Each slides support can contain 100 microscope slides, easily extractable.
- Each cavity is numbered.
- The transparent lids guarantee an easy check of the contents, protecting the slides from dust and other foreign bodies.
Specifications
4 | |
18.7 in. | |
700 mm | |
19.3 in. | |
18.4 kg | |
475 mm | |
490 mm | |
PMMA/SS |
Cabinet | |
Slide Storage | |
27.6 in. | |
18.7 in. | |
40.5 lb. | |
475 mm | |
Transparent |