Epredia™ SlideMate™ AS Slide Printer
Reduce specimen identification errors while enhancing lean workflow with the Epredia™ SlideMate™ AS On-Demand Slide Printer.
Supplier:  Epredia™ A83900005
 View more versions of this product
Catalog No. A83900005
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The SlideMate AS is the next generation Epredia Slide Printer delivering a smaller footprint than previous models, making it even easier to fit next to the microtome. The SlideMate AS promotes single piece workflow though unique features designed specifically for on-demand printing.
  • Fully integrated color touch-screen and simple software are easy to use
  • Build-in barcode scanner reads cassette information and imports the data to the slide label
  • Connects easily to other computers or Laboratory Information Systems
  • Prints directly onto the slide, eliminating the need for hand writing or labels
Specifications
Specifications
SlideMate AS On-Demand Printing System
SlideMate AS on-demand printer with slide delivery system, color touch-screen, built-in scanner, two USB ports, one ethernet port, power cord and advanced software enabling label creation, special panel printing and import from LIS