missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ SlideMate™ Adhesion Microscope Slides
SlideMate™ Adhesion Microscope Slides offers crisp, clean printing. Designed for use on SlideMate thermal printers. Reduces identification issues common to handwritten labels when paired with SlideMate printers. Comes 144 slides per gross; 10 gross per case.
$121.13 - $1210.90
Specifications
Edges | Ground |
---|---|
Thickness (Metric) | 1 mm |
Length (Metric) | 75 mm |
Width (Metric) | 25 mm |
Coating | Adhesion |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
22042938
|
Epredia™
TT5041IPS82181 |
Blue |
Case of 1440 Each for $1,210.90
|
|
|||||
22042949
|
Epredia™
TT8041IPS82181 |
Yellow |
Case of 1440 Each for $1,210.90
|
|
|||||
22042951
|
Epredia™
TT6041IPS82181 |
Green |
Case of 1440 Each for $1,210.90
|
|
|||||
22-042-936
|
Epredia™
TT4041IPS82181 |
White |
Pack of 144 Each for $121.13
Case of 10 Pack for $1,211.30
|
|
|||||
Description
- Manufactured in the USA; SlideMate slides are smooth, three-quarter inch white, blue, green, pink or yellow tab slides validated to work with the SlideMate AS slide printer.
- The tab is impervious to virtually all common laboratory chemicals and reagents.
- Available in adhesion and non-adhesion formulas. SlideMate Plus Adhesion Slides feature adhesion coating for routine clinical applications.
- Epredia SlideMate slides are made from extra-white glass, manufactured to high quality standards. The clarity achieved during the manufacturing process helps ensure high levels of transparency to aid in accurate diagnoses.
- SlideMate slides come pre-cleaned, flat with finely ground edges. Each package comes wrapped to help protect against humidity and moisture. The packaging is also designed to assist with loading the slides into SlideMate printer hoppers to prevent contamination due to handling.
Specifications
Ground | |
75 mm | |
Adhesion | |
75 x 25 mm |
1 mm | |
25 mm | |
90° |