Thermo Scientific™ Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for four inner-door, ultra-low temperature freezers.
Supplier:  Thermo Scientific™ 1950520
 View more versions of this product
Catalog No. 1950520
Explore available promotions
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 4 shelf Series Freezers as well as well as previous generation Forma™ 88000 and TSU Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
Specifications
Specifications
Side Access Freezer Rack (Adjustable)
Silver
Stainless Steel
5
13.5 cm
29.5 cm
26.8 in.
2 in. Box Racks - Adjustable side access rack
Boxes (2 in.)
25 Boxes/Rack
Thermo Scientific TDE Series and TSX Series (4 shelves)
5.3 in.
11.6 in.
68.1 cm