Thermo Scientific™ Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for four inner-door, ultra-low temperature freezers.
Supplier:  Thermo Scientific™ 920091
 View more versions of this product
Catalog No. 920091
Explore available promotions
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 4 shelf Series Freezers as well as well as previous generation Forma™ 88000 and TSU Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
Specifications
Specifications
Box Racks - Sliding Drawer
Silver
Stainless Steel
Square
14 cm
30.2 cm
25.5 in.
3 in. Box Rack - Sliding Drawer
Boxes (3 in.)
15 Boxes/Rack
Thermo Scientific TDE Series and TSX Series (4 shelves)
5.5 in.
11.9 in.
64.7 cm