Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
Supplier:  Thermo Scientific™ 398323
 View more versions of this product
Catalog No. 398323
Explore available promotions
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
Specifications
Specifications
Side Access Freezer Rack (Microplate/Locking Rod)
Silver
Stainless Steel
Rectangular
14 cm
23.9 cm
22 in.
Side Access Microplate Rack w/Locking Rod - Standard or Deepwell
Microplates with Lids
84 Plates/Rack
17.3 and 23 cu. ft. capacity upright freezers
5.5 in.
9.4 in.
55.9 cm