Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
Supplier:  Thermo Scientific™ 398325
 View more versions of this product
Catalog No. 398325
Explore available promotions
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
Specifications
Specifications
Side Access Freezer Rack (Adjustable)
Silver
Stainless Steel
3 Tiers
13.7 cm
23.9 cm
16.1 in.
3 in. Box Racks - Adjustable side access rack
Boxes (3 in.)
9 Boxes/Rack
13 cu. ft. capacity upright freezers
5.4 in.
9.4 in.
41.9 cm