Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
Supplier:  Thermo Scientific™ 398330
 View more versions of this product
Catalog No. 398330
Explore available promotions
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
Specifications
Specifications
Box Racks - Sliding Drawer
Silver
Stainless Steel
Rectangular
14 cm
24 cm
22 in.
3 in. Box Rack - Sliding Drawer
Boxes (3 in.)
12 Boxes/Rack
17.3 and 23 cu. ft. capacity upright freezers
5.5 in.
9.5 in.
55.9 cm