Greiner Bio-One SILVERseal™ Adhesive Microplate Sealer
Pierceable and coated with acrylate adhesive. Greiner Bio-One SILVERseal™ Adhesive Microplate Sealer is robust, heat- resistant and with pierceable aluminum film.
Supplier:  Greiner Bio-One 676090
Catalog No. 07000379
Explore available promotions
Description
Description
  • Coated with acrylate adhesive
  • Non-Pyrogenic
  • Temperature-resistant from -80°C to +110°C
  • Ideal for the storage of sample material and active agents
  • Free of detectable DNase, RNase, human DNA
  • Ideal for
Specifications
Specifications
100
Sealer
Yes
PCR
Acrylate
Microplate
Videos
SDS
Documents
Documents
Promotions
Promotions
For research use only. Not for use in diagnostic procedures, unless otherwise indicated.