missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whirl-Pak™ Sludge Judge Ultra™ Sample Pole Unit and Components

Made from rigid plastic tubing in three 5 ft. (1.53 m) sections, this unit and complementing replacement sections provide maximum strength for liquid sampling at long distances.

$158.68 - $541.00

Specifications

Certifications/Compliance ISO 9001 and ISO 13485
Volume (Metric) 89 mL/ft.
For Use With (Application) Sludge Judge Ultra
For Use With (Equipment) Sludge Judge Ultra
Graduated Yes
View More Specs

Products 4
Catalog Number Mfr. No. Type Includes Price Quantity  
Catalog Number Mfr. No. Type Includes Price Quantity  
0181296
View Documents
Whirl-Pak™
B01387
Sludge Judge Ultra Top Section Top Section, Rope
Each for $158.68
 
01-812-97
View Documents
Whirl-Pak™
B01388
Sludge Judge Ultra Extension Section Middle Section
Each for $167.39
 
01-812-98
View Documents
Whirl-Pak™
B01389
Sludge Judge Ultra Bottom Section Bottom Section, Valve
Each for $251.47
 
01-812-95
View Documents
Whirl-Pak™
B01386
Sludge Judge Ultra Complete Unit All Sections
Each for $541.00
 
Description

Description

Made from rigid plastic tubing in three 5 ft. (1.53m) sections, provides maximum strength for liquid sampling at long distances

  • Unit 01-812-95 is designed for sampling 5% or less solids.
  • Constructed of extremely strong polycarbonate that is treated with an ultraviolet stabilizer to help reduce deterioration from the sun harmful rays.
  • Material is very rigid, minimizing bending of the unit when it is full of water or other liquids.
  • Durable in cold temperatures, can withstand heat up to 280°F (138°C) with careful handling.
  • The 0.75 in. (1.9 cm) diameter tubing is marked with blue tape to designate 1 ft. (0.31 m) measurements on the tubing.
  • Unit is made up of three 5 ft. (1.53 m) sections (top, extension, bottom). Individual sections can be combined as required.
  • Not autoclavable.
  • Replacement parts for Sludge Judge; Ultra Sample Pole part number 01-812-95 include extension section, top section with rope, and/or bottom section with valve.
Specifications

Specifications

ISO 9001 and ISO 13485
Sludge Judge Ultra
Yes
1.53 m
89 mL/ft.
Sludge Judge Ultra
5 ft.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'