missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One Small Volume™ 384 Well HiBase Polypropylene Microplates

Reduces dead volume. Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates are specifically designed to accommodate heat-sealing systems.

Supplier:  Greiner Bio-One 784900

Catalog No. 07000916


Explore available promotions

Description

Description

  • Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
  • Long-term surface stability without degradation or leaching
  • Higher assay sensitivity with reduced background
  • Good signal to noise ratio
  • Fre
Specifications

Specifications

Alphanumeric
16 x 24
Inner packs of 10
Polystyrene
384
Protein-Repellent Non-binding
Flat Bottom (F-well)
-20°C to +60°C
Standard
Not Autoclavable
Black
4 to 25 μL
Non-sterile
28 μL
40/Cs.
Black
Videos
SDS
Documents

Documents

Promotions

Promotions