Learn More
Thermo Scientific™ SMART Digest™ RNase Kits
Use SMART Digest™ RNase Kits in your oligonucleotide mapping and quantitation workflows to obtain high quality analytical results.
$5473.65 - $6187.62
Specifications
Format | Bulk Magnetic SMART Digest Resin |
---|---|
For Use With (Application) | Oligonucleotide Mapping |
Product Line | SMART Digest |
Type | RNA Digestion |
Includes | Bulk Magnetic SMART Digest Resin, SMART Digest Buffer |
Catalog Number | Mfr. No. | Cleavage | Enzyme | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Cleavage | Enzyme | Price | Quantity | |||||
01-257-521
|
Thermo Scientific™
60120101 |
Degrades single-stranded RNA at G residues | RNase T1 |
Each for $6,187.62
|
|
|||||
01-257-522
|
Thermo Scientific™
60120102 |
Degrades single-stranded RNA at C and U residues | RNase A |
Each for $5,473.65
|
|
|||||
Description
Obtain high-quality analytical results from mRNA digestions using Thermo Scientific™ SMART Digest™ RNase Kits. A significant advance in sample preparation for oligonucleotide mapping and quantitation, these kits provide fast and simple mRNA digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that is compatible with automation.
These RNase kits use optimized, heat stable enzymes on magnetic beads, allowing for control of partial digestion to produce required fragments of ideal length for sequencing by HRAM-MS.
Easy and Simple
- A simple process and control of digestion time ensure required fragment length for comprehensive mRNA sequencing by HRAM-MS
- Easily automate the entire protocol for high-throughput processing
- Single nuclease digestion allows full characterization of mRNA
Reliable Results
- Increased reproducibility over existing protocols
- Higher data confidence
- No nuclease contamination of the analytical column
RNase kits are available with with two endoribonuclease enzyme specificities:
- RNase T1 degrades single-stranded RNA at G residues
- RNase A degrades single-stranded RNA at C and U residues
Specifications
Bulk Magnetic SMART Digest Resin | |
SMART Digest | |
Bulk Magnetic SMART Digest Resin, SMART Digest Buffer |
Oligonucleotide Mapping | |
RNA Digestion |