MilliporeSigma™ Supelco™ Smart SPME Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.
Supplier:  MilliporeSigma™ Supelco™ 548650U
 View more versions of this product
Catalog No. 111023069
Explore available promotions
Description
Description
  • Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.
  • Each Smart fiber assembly, specifically designed to be compatible with the PAL 3, Series II Smart autosampler, incorporates a SPME fiber within a holder and a programmable computer chip coded with fiber chemistry type, dimensions, and lot number.
  • The Smart chip provides traceability by recording fiber stroke count, injection and conditioning durations, usage dates, and expiration date.
  • The one piece fiber/holder design makes the Smart SPME assembly easier to handle and install than traditional SPME.
  • In addition, compatibility with the SPME tool on the PAL 3 autosampler allows for more efficient method development and completely automated sample preparation, thus increasing overall productivity.
Specifications
Specifications
PAL 3, Series II Smart Autosampler
Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
65 μm
Fiber Assembly
23 G
Orange/Red