missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wanzl SmartCell™ Starter Unit with Baskets

SmartCell™ Shelving delivers industry-leading open wire storage that allows for easy identification of materials. Our quickly adjustable storage compartments increase the efficiency of both materials and staff management alike.

$1515.40 - $5288.49

Specifications

For Use With (Application) Shelving
For Use With (Equipment) SmartCell™
Shelf Type Starter Unit
Width (English) 74 in.
Color Chrome
View More Specs

Products 12
Catalog Number Mfr. No. Length (English) Includes Price Quantity  
Catalog Number Mfr. No. Length (English) Includes Price Quantity  
11222002
View Documents
Wanzl
THSC186S
18 in. 6 Baskets
Each for $1,515.40
 
11222004
View Documents
Wanzl
THSC366S
36 in. 6 Baskets
Each for $2,177.17
 
11222006
View Documents
Wanzl
THSC486S
48 in. 6 Baskets
Each for $2,518.81
 
11222014
View Documents
Wanzl
THSC187S
18 in. 7 Baskets
Each for $1,656.74
 
11222016
View Documents
Wanzl
THSC367S
36 in. 7 Baskets
Each for $2,410.23
 
11222018
View Documents
Wanzl
THSC487S
48 in. 7 Baskets
Each for $2,799.62
 
11222008
View Documents
Wanzl
THSC186S2
18 in. 12 Baskets
Each for $2,873.57
 
11222010
View Documents
Wanzl
THSC366S2
36 in. 12 Baskets
Each for $4,143.74
 
11222012
View Documents
Wanzl
THSC486S2
48 in. 12 Baskets
Each for $4,734.36
 
11222020
View Documents
Wanzl
THSC187S2
18 in. 14 Baskets
Each for $3,153.43
 
11222022
View Documents
Wanzl
THSC367S2
36 in. 14 Baskets
Each for $4,610.81
 
11222024
View Documents
Wanzl
THSC487S2
48 in. 14 Baskets
Each for $5,288.49
 
Description

Description

  • Fine mesh wire baskets come in various sizes and are suitable for mounting on louvered panels or cantilever pedestal frames.
  • Adjustable dividers provide endless variations to storage space and also create opportunities to implement lean inventory management practices, such as Kanban and PAR level systems.
  • All baskets are plated in hospital-grade chrome and the framework is finished with industrial strength powder coat to enhance the durability and longevity of the system.
  • Bottom dust shelf/cover provides a barrier and protection to bottom basket contents, maintaining Joint Commission standards.
  • Wide variety of accessories available to customize the system to your application
Specifications

Specifications

Shelving
Starter Unit
Chrome
SmartCell™
74 in.
Steel
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'