missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 3cc/70.5IU Kits
Only available in the USA and selectively in Canada. Pro-Vent™ Plus 3 mL Arterial Blood Sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results. The syringes are available with a broad choice of product features.
$1112.56 - $1112.56
Specifications
For Use With (Application) | Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results |
---|---|
Volume (Metric) | 3 mL |
Sterility | Sterile |
Catalog Number | Mfr. No. | Product Type | Quantity | Includes | Packaging | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Includes | Packaging | Price | Quantity | ||
22004179
|
ICU Medical
CIA1074645 |
Arterial Blood Sampling Kit | 200/Cs. | 3mL luer-slip syringe with Filter-Pro™ device, 23G x 1 in. preattached needle with Needle-Pro™ needle safety device, adhesive bandage, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant | Sterile Tyvek™ |
Case of 200 Each for $1,112.56
|
|
||
Description
- Advanced-formula dry lithium heparin
- Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
- Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
- Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
- Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
- Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
- Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Order Info
Sterile Tyvek packaging except as noted.
†Polybagged.
Specifications
Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results | |
3 mL | |
Sterile |