ICU Medical Jelco™ Saf-T Holder™ Blood Culture Holders and Transfer Devices
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
Supplier: ICU Medical 96005
Description
- Eliminate potential exposures and holder reuse
- Provide safe needleless transfer after a syringe draw
Specifications
Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw | |
Saf-T Holder™ Blood Culture Device, with female luer |