missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Guardian™ CP-14, Chemical Resistant Smooth Finish 14 mil Butyl Gloves
Medium weight butyl gloves with fitted hand styling and a smooth finish offer protection against a wide range of hazardous chemicals.
$67.41 - $67.41
Specifications
Certifications/Compliance | ASTM F739-05 |
---|---|
Certifications/Compliance (Europe) | CE |
Color | Black |
Hand | Specific |
Latex Free | Yes |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19321104
|
Guardian™
GGL51401 |
X-Small |
Pair for $67.41
|
|
|||||
19321105
|
Guardian™
51402 |
Small |
Pair for $67.41
|
|
|||||
19321106
|
Guardian™
51403 |
Medium |
Pair for $67.41
|
|
|||||
19321107
|
Guardian™
51404 |
Large |
Pair for $67.41
|
|
|||||
19321108
|
Guardian™
51405 |
X-Large |
Pair for $67.41
|
|
|||||
Description
- Butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with smooth texture
- Protects against wide range of hazardous chemicals
- 14 mil thickness, 14 in. length
- Made in the USA
Specifications
ASTM F739-05 | |
Black | |
Yes | |
Powdered | |
0.35 mm | |
Gauntlet/Rolled | |
Unlined | |
14 in. |
CE | |
Specific | |
Butyl | |
14 mil | |
Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones | |
Smooth | |
ISO 9001 | |
35.5 cm |