Guardian™ CP-25, Chemical Resistant Smooth Finish 25 mil Butyl Gloves
Heavyweight butyl gloves with fitted hand styling and a smooth finish offer protection against a wide range of hazardous chemicals.
Supplier:  Guardian™ 52503
 View more versions of this product
Catalog No. 19321111
Explore available promotions
Description
Description
  • Butyl material resists oxidation, ozone corrosion and abrasions
  • Reusable and easy to clean
  • Fitted hand style with smooth texture
  • Protects against wide range of hazardous chemicals
  • 25 mil thickness, 14 in. length
  • Made in the USA
Specifications
Specifications
ASTM F739-05
Black
Yes
Powdered
25 mil
Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones
Smooth
ISO 9001
14 in.
CE
Specific
Butyl
Medium
0.63 mm
Gauntlet/Rolled
Unlined
52503
35.5 cm