Motic™ SMZ 171 Stereo Microscopes
The SMZ-171 model terminates Motic™â€™s SMZ stereo microscope line by using modern ESD-resistant materials and optimized LED illuminations for a wide range of biomedical and material science applications.
Supplier:  Motic™ 1100200600754
 View more versions of this product
Catalog No. 01-258-017
Explore available promotions
Description
Description
The SMZ-171 is an advanced stereo microscope for biomedical work and industrial quality control.
  • Greenough Zoom Optical System
  • 45° stereomicroscope head
  • 6.7:1 Zoom Ratio
  • Diopter Adjustment: ±5
Specifications
Specifications
LED Stand
Auxiliary Objectives (0.3X, 0.5X, 0.63X, 0.75X, 1.5X, 2X)
LED Transmitted Illumination
110 mm
10X
Binocular
WF 10X/23 mm