missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ SnakeSkin™ Dialysis Tubing

Perform desalting and buffer exchange for 10 to 100 mL samples with this 3.5K, 7K, and 10K molecular weight cut-off (MWCO), ready-to-use form of traditional dialysis membrane tubing. Securely close dialysis tubing up to 50 mm flat diameter with the sturdy, non-floating tube SnakeSkin Dialysis Clips.

Supplier:  Thermo Scientific™ 68011

 View more versions of this product

Catalog No. PI68011


Explore available promotions

Includes: This item, PI68011, is the Dialysis Clips only.

Description

Description

The SnakeSkin 3.5K, 7K, and 10K MWCO dialysis tubing is an easy and ready-to-use traditional dialysis membrane tubing that allows desalting and buffer exchange for samples of 10 to 100 mL. SnakeSkin Dialysis Tubing Clips are sturdy, non-floating tube closures recommended for use with SnakeSkin Dialysis Tubing, but are suitable for any dialysis tubing with flat tube diameters up to 50 mm.

The easy-open, pleated format of SnakeSkin Dialysis Tubing was developed to simplify large volume sample dialysis. To use, simply pull out and cut off the required length of tubing, fold over one end of tubing and apply a dialysis clip to close it, then add sample at the open end and use a second clip to close the remaining end. Clips are sold separately. In comparison, many traditional dialysis tubing membranes are supplied as dry, flattened rolls that are difficult to open and often require extensive soaking before they can be used.

The SnakeSkin membrane-pleating process does not change the tubing's MWCO. Also, any low MW contaminants present are removed during the dialysis process. SnakeSkin Dialysis Tubing is made from the same type of regenerated cellulose as flat tubing, so it matches the dialysis performance of conventional tubing.

Features of SnakeSkin Dialysis Tubing:
Convenient—ready to use, pre-wetted, pleated tube
High recovery—greater than 90% protein recovery
Speed—dialysis is generally completed in 4–6 hours
Stability—compatible with a variety of laboratory solutions, including acids, bases, hydrophobic solvents, and alcohols

Features of SnakeSkin Dialysis Tubing Clips:
Durable—polyamide composition for long-life
Dove-tail latch—prevent sample loss
Heat resistant—autoclave up to 134°C

SnakeSkin Dialysis Tubing, 3.5K or 10K MWCO, is composed of regenerated-cellulose and supplied as an open, pleated (telescoped) tube. It is supplied in 20-cm sticks containing 35 feet of dialysis tubing having a 16-, 22-, or 35-mm circular internal diameter (I.D.). With similar capacity to 10.5 meters of 23–50 mm diameter dry flat width tubing, the hydrated SnakeSkin Dialysis Tubing holds ∼2–10 mL of sample per centimeter of length.

SnakeSkin 7K MWCO dialysis tubing is supplied in 20-cm sticks containing 35 feet of dialysis tubing having a 22-mm circular internal diameter (I.D.). Equivalent in capacity to 10.5 meters of 34-mm diameter dry flat width tubing, the hydrated SnakeSkin Dialysis Tubing holds ∼3.7 mL of sample per centimeter of length.

3.5K MWCO tubing specifications:
Membrane type: regenerated cellulose
Tubing nominal dry thickness: 0.9–1.2 mil (22–30 μm)
Glycerol content: trace
Sulfur content: 0.1–0.15%
Heavy metals content: trace

7K MWCO tubing specifications:
Membrane type: regenerated cellulose
Tubing nominal dry thickness: 0.9–1.2 mil (22–30 μm)
Glycerol content: 13%
Sulfur content: 0.1–0.15%
Heavy metals content: trace

10K MWCO tubing specifications:
Membrane type: regenerated cellulose
Tubing nominal dry thickness: 0.9–1.2 mil (22–30 μm)
Glycerol content: 21%
Sulfur content: 0.1–0.15%
Heavy metals content: trace

Dialysis clip specifications:
Height: 10 mm
Width: 16 mm
Inner Length: 50 mm (maximum closure width)
Outer Length: 66 mm (end to end)
Composition: polyamide (nylon) derivative

Specifications

Specifications

Dialysis Clips
Not Applicable
Not Applicable
SnakeSkin™
Polyamide
6 Clips
Videos
SDS
Documents

Documents

Promotions

Promotions