Learn More
Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes
Improve sample recovery by significantly reducing surface binding with these tubes that do not have additives or coatings.
$107.38 - $219.07
Catalog Number | Mfr. No. | Description | Color | Max. RCF | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Color | Max. RCF | Price | Quantity | |||
21-402-905
|
Thermo Scientific™
3453 |
2.0mL Graduated, Sterile | Natural | 25,000 x g |
Pack of 500 Each for $125.48
Case of 10 Pack for $1,254.80
|
|
|||
21-402-904
|
Thermo Scientific™
3434 |
2.0mL Graduated, Nonsterile | Natural | 25,000 x g |
Pack of 500 Each for $139.79
Case of 10 Pack for $1,397.90
|
|
|||
21-402-903
![]() |
Thermo Scientific™
3451 |
1.5mL Graduated, Sterile | Natural | 26,000 x g |
Pack of 500 Each for $107.38
Case of 10 Pack for $1,073.80
|
![]() |
|||
21-403-189
|
Thermo Scientific™
3446 |
0.6mL Graduated, Nonsterile | Natural | 30,000 x g |
Pack of 1000 Each for $198.90
Case of 10 Pack for $1,989.00
|
|
|||
21-403-190
|
Thermo Scientific™
3449 |
0.6mL Graduated, Sterile | Natural | 30,000 x g |
Pack of 1000 Each for $219.07
Case of 10 Pack for $2,190.70
|
|
|||
Description
Improve sample recovery by significantly reducing surface binding with Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes. Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. With no additives or coatings, these tubes — available in 0.6mL, 1.5mL and 2.0mL sizes — offer maximum sample recovery by significantly reducing surface binding.
- The duckbill cap reduces the pressure required to open the tube with a wider contact surface, yet ensures the cap will remain closed during centrifugation
- Low retention properties are not achieved by a coating or plastic-embedding processes, ensuring that samples will not be contaminated from these additives
- Tubes will remain closed during centrifugation yet open easily with one hand as needed
- Tubes are available in amber to protect your light-sensitive samples
- Graduation marks on the microcentrifuge tube allow for effective sample reference
- A variety of packaging options are available to help with contamination control in the lab
- For stringent sterility requirements, individually wrapped sterile tubes are available
- Color options, in addition to natural and amber, are red, blue, green, yellow and violet and a mixed color packaging
- Manufactured from virgin polypropylene, these microcentrifuge tubes ensure sample integrity during boiling, freezing and centrifugation
- Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
- Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Recommended for:
Daily laboratory usage including centrifugation and short-term sample storage
Specifications
Flat | |
Natural | |
Polypropylene | |
Yes |