missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube

Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.

Supplier:  Eppendorf™ 022363646

 View more versions of this product

Catalog No. 05-402-20


Explore available promotions

Description

Description

  • The hinged Eppendorf Safe-Lock lid prevents unintended opening during incubation and storage.
  • g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss in the event of tube breakage and providing enhanced safety when handling hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides, substances known to leach from plastic consumables into the sample and adversely impact bio-assay results.
  • Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures.
  • Precise lid sealing minimizes evaporation rates during long-term storage.
  • The large frosted lid and side surface facilitate easy labeling.
  • Ensured functionality in a temperature range from -86 °C to 100 °C.
  • Autoclavable when open (121°C, 20 min.) for effective sterilization.
  • Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality, ensuring tested purity.

Order Info

Assorted packages include 100 each of natural, blue, green, red and yellow

Specifications

Specifications

Polypropylene
0.5 mL
33 mm
500 Pk.
-122.8°F to +212°F
Non-sterile
Yes
Safe-lock Lid
1.3 in.
30,000 x g
0.25 mL
Autoclavable
Blue
Polypropylene
500 Tubes (1 Bag x 500 Tubes)
-86°C to +100°C
Eppendorf Quality (Non-sterile)
0.02 oz.
Cylindrical
Conical
Locking Snap-cap Microtube
9 mm
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions