missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Supelco™ Snap Ring Polypropylene Autosampler Microvial
Autoclavable, easy to maintain. A vial is used to store or as a closure type.
$34.50 - $276.01
Catalog Number | Mfr. No. | No. per Pack | Volume (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. per Pack | Volume (Metric) | Price | Quantity | ||||
111010037
|
MilliporeSigma™ Supelco™
27254 |
1000 | 0.75 mL |
Each for $276.01
|
|
||||
111011843
|
MilliporeSigma™ Supelco™
27423 |
100 | 0.1 mL |
Each for $53.50
|
|
||||
111011968
|
MilliporeSigma™ Supelco™
27422 |
100 | 0.5 mL |
Each for $41.10
|
|
||||
111012028
|
MilliporeSigma™ Supelco™
27253 |
100 | 0.75 mL |
Each for $34.50
|
|
||||
Specifications
Natural Polypropylene | |
Large opening, (6 mm) |