Socorex Self-Refilling Repetitive Syringe from DWK Life Sciences™
Allows repetitive delivery of preset aliquots from a single reservoir via silicone tubing
Supplier:  Socorex 851371
 View more versions of this product
Catalog No. 13-689-50A
Explore available promotions
Includes: Blunt-end cannula with Luer-Lok™ hub, long and short chrome-plated intake/air vent cannula, sinker, and silicone tubing
Description
Description
Semiautomatic syringe consists of a spring-loaded, stainless-steel plunger and replaceable ground-glass barrel. Five adjustable models handle volumes from 0.05 to 20mL.
  • Detachable three-way valve with corrosion-proof springs and stainless-steel check valve balls ensures accuracy of ±1%
  • For additional corrosion protection, ground-glass barrel is supplied with Viton™ washers and corrosion-resistant stainless-steel plunger
  • Autoclavable
Order Info
To convert unit to a multichannel syringe, attach a Microtest Manifold. For replacement parts, call your Fisher Scientific™ Customer Service Representative.
Specifications
Specifications
±1%
1
0 to 1 mL
0.05mL
Autoclavable
Self-Refilling Repetitive Syringe
Blunt end cannula with luer lock hub, long and short chrome plated intake/air vent cannula, sinker and Silicone tubing