Learn More
Soda lime, indicator grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1935236
Description
Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSoda lime′s ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.
Solubility
Soluble in water.
Notes
Air sensitive and hygroscopic. Incompatible with acids, water, fluorine, chloroform and trichloroethylene. It is a noncombustible material. Generates heat on hydrolysis.
Specifications
Soda lime | |
500 g | |
MFCD00134094 | |
14,8569 | |
Soluble in water. | |
[OH-].O=[Ca].[Na+] | |
96.074 | |
Odorless |
8006-28-8 | |
CaHNaO2 | |
UN1907 | |
soda lime, soda-lime, sodasorb, soda lime nf, sodium hydroxide na oh , mixt. with lime, soda-lime, granular, soda lime, indicating, soda lime, indicator grade, soda lime with sodium hydroxide, soda lime, indicating 8-12 mesh | |
HUAUNKAZQWMVFY-UHFFFAOYSA-M | |
sodium;oxocalcium;hydroxide | |
66545795 | |
Air sensitive; Hygroscopic |
Chemical Identifiers
8006-28-8 | |
96.074 | |
HUAUNKAZQWMVFY-UHFFFAOYSA-M | |
66545795 | |
[OH-].O=[Ca].[Na+] |
CaHNaO2 | |
MFCD00134094 | |
soda lime, soda-lime, sodasorb, soda lime nf, sodium hydroxide na oh , mixt. with lime, soda-lime, granular, soda lime, indicating, soda lime, indicator grade, soda lime with sodium hydroxide, soda lime, indicating 8-12 mesh | |
sodium;oxocalcium;hydroxide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: SODA LIME
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only