missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 1-heptanesulfonate, 99% (dry wt.), water <2%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1091722

 View more versions of this product

Catalog No. AAA1091722


Explore available promotions

Description

Description

HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium 1-heptanesulfonate
22767-50-6
Odorless
C7H15NaO3S
100 g
sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
REFMEZARFCPESH-UHFFFAOYSA-M
sodium;heptane-1-sulfonate
23672332
99%
Water <2%
>300°C
(dry wt.)
MFCD00007543
3731762
Soluble in water.
CCCCCCCS(=O)(=O)[O-].[Na+]
202.244
202.24

Chemical Identifiers

22767-50-6
202.244
REFMEZARFCPESH-UHFFFAOYSA-M
23672332
CCCCCCCS(=O)(=O)[O-].[Na+]
C7H15NaO3S
MFCD00007543
sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
sodium;heptane-1-sulfonate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 245-210-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only