Learn More
Sodium 1-hexanesulfonate, 99%, Thermo Scientific Chemicals
$81.07 - $753.66
Chemical Identifiers
CAS | 2832-45-3 |
---|---|
Molecular Formula | C6H13NaO3S |
Molecular Weight (g/mol) | 188.22 |
MDL Number | MFCD00007542 |
InChI Key | QWSZRRAAFHGKCH-UHFFFAOYSA-M |
Synonym | sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt |
PubChem CID | 23677630 |
IUPAC Name | sodium;hexane-1-sulfonate |
SMILES | [Na+].CCCCCCS([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1327806
|
Thermo Scientific Chemicals
A1327806 |
5 g |
Each for $81.07
|
|
|||||
AAA1327814
|
Thermo Scientific Chemicals
A1327814 |
25 g |
Each for $236.89
|
|
|||||
AAA1327822
|
Thermo Scientific Chemicals
A1327822 |
100 g |
Each for $753.66
|
|
|||||
Description
Sodium 1-hexanesulfonate is used as a base in the organic coupling reactions such as Suzuki coupling reactions. It is used as an ion pairing reagent for HPLC and involved in the analysis of peptides and proteins. It is also used in high performance capillary electrophoresis analysis of peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium 1-hexanesulfonate is used as a base in the organic coupling reactions such as Suzuki coupling reactions. It is used as an ion pairing reagent for HPLC and involved in the analysis of peptides and proteins. It is also used in high performance capillary electrophoresis analysis of peptides.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
2832-45-3 | |
188.22 | |
QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
23677630 | |
[Na+].CCCCCCS([O-])(=O)=O |
C6H13NaO3S | |
MFCD00007542 | |
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
sodium;hexane-1-sulfonate |
Specifications
2832-45-3 | |
99% | |
MFCD00007542 | |
3727014 | |
Soluble in water. | |
[Na+].CCCCCCS([O-])(=O)=O | |
188.22 | |
188.22 | |
Powder |
>300°C | |
C6H13NaO3S | |
5 g | |
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
sodium;hexane-1-sulfonate | |
23677630 | |
99% | |
Sodium 1-hexanesulfonate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-601-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only