missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium 1-Naphthol-8-sulfonate 96.0+%, TCI America™
$29.41 - $278.54
Chemical Identifiers
CAS | 20215-36-5 |
---|---|
Molecular Formula | C10H7NaO4S |
Molecular Weight (g/mol) | 246.212 |
MDL Number | MFCD00021524 |
InChI Key | PEYXCGCCIWJCTM-UHFFFAOYSA-M |
Synonym | 8-Hydroxy-1-naphthalenesulfonic Acid Sodium Salt, Sodium 8-Hydroxy-1-naphthalenesulfonate, 1-Naphthol-8-sulfonic Acid Sodium Salt |
PubChem CID | 23672328 |
IUPAC Name | sodium;8-hydroxynaphthalene-1-sulfonate |
SMILES | C1=CC2=C(C(=C1)O)C(=CC=C2)S(=O)(=O)[O-].[Na+] |
Chemical Identifiers
20215-36-5 | |
246.212 | |
PEYXCGCCIWJCTM-UHFFFAOYSA-M | |
23672328 | |
C1=CC2=C(C(=C1)O)C(=CC=C2)S(=O)(=O)[O-].[Na+] |
C10H7NaO4S | |
MFCD00021524 | |
8-Hydroxy-1-naphthalenesulfonic Acid Sodium Salt, Sodium 8-Hydroxy-1-naphthalenesulfonate, 1-Naphthol-8-sulfonic Acid Sodium Salt | |
sodium;8-hydroxynaphthalene-1-sulfonate |
Specifications
20215-36-5 | |
MFCD00021524 | |
8-Hydroxy-1-naphthalenesulfonic Acid Sodium Salt, Sodium 8-Hydroxy-1-naphthalenesulfonate, 1-Naphthol-8-sulfonic Acid Sodium Salt | |
C1=CC2=C(C(=C1)O)C(=CC=C2)S(=O)(=O)[O-].[Na+] | |
246.212 | |
246.21 | |
Crystalline Powder |
C10H7NaO4S | |
25 g | |
PEYXCGCCIWJCTM-UHFFFAOYSA-M | |
sodium;8-hydroxynaphthalene-1-sulfonate | |
23672328 | |
≥96.0% (HPLC) | |
Sodium 1-Naphthol-8-sulfonate |
Safety and Handling
EINECSNumber : (4)-0477
TSCA : No