missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals

$152.32 - $549.68

Chemical Identifiers

CAS 35192-74-6
Molecular Formula C9H19NaO3S
Molecular Weight (g/mol) 230.298
MDL Number MFCD00025010
InChI Key RUYRDULZOKULPK-UHFFFAOYSA-M
Synonym sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate
PubChem CID 23664777
IUPAC Name sodium;nonane-1-sulfonate
SMILES CCCCCCCCCS(=O)(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0640706
View Documents
Thermo Scientific Chemicals
L0640706
5 g N/A
 
AAL0640714
View Documents
Thermo Scientific Chemicals
L0640714
25 g N/A
 
Description

Description

Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

35192-74-6
230.298
RUYRDULZOKULPK-UHFFFAOYSA-M
23664777
CCCCCCCCCS(=O)(=O)[O-].[Na+]
C9H19NaO3S
MFCD00025010
sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate
sodium;nonane-1-sulfonate

Specifications

35192-74-6
C9H19NaO3S
MFCD00025010
3738676
Soluble in water.
CCCCCCCCCS(=O)(=O)[O-].[Na+]
230.298
230.3
Sodium 1-nonanesulfonate
>300°C
CH3(CH2)8SO3Na
5 g
sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate
RUYRDULZOKULPK-UHFFFAOYSA-M
sodium;nonane-1-sulfonate
23664777
≥98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only