Learn More
Sodium 1-octanesulfonate hydrate, HPLC grade, 99+%, Thermo Scientific Chemicals
Ion-pair reagent for HPLC analysis
Supplier: Thermo Scientific Chemicals 04263614
Description
Ion-pair reagent for HPLC analysisSodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium 1-octanesulfonate hydrate | |
>300°C | |
MFCD00007544 | |
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8 | |
HRQDCDQDOPSGBR-UHFFFAOYSA-M | |
sodium;octane-1-sulfonate | |
23669624 | |
≥99% | |
Crystalline |
5324-84-5 | |
C8H17NaO3S | |
25 g | |
Soluble in water producing a clear and colorless solution. | |
[Na+].CCCCCCCCS([O-])(=O)=O | |
216.27 | |
216.28 (Anhydrous) | |
HPLC |
Chemical Identifiers
5324-84-5 | |
216.27 | |
HRQDCDQDOPSGBR-UHFFFAOYSA-M | |
23669624 | |
[Na+].CCCCCCCCS([O-])(=O)=O |
C8H17NaO3S | |
MFCD00007544 | |
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8 | |
sodium;octane-1-sulfonate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 226-195-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only