missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 1-octanesulfonate hydrate, HPLC grade, 99+%, Thermo Scientific Chemicals

Ion-pair reagent for HPLC analysis

Supplier:  Thermo Scientific Chemicals 04263622

 View more versions of this product

Catalog No. AA4263622


Explore available promotions

Description

Description

Ion-pair reagent for HPLC analysisSodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium 1-octanesulfonate hydrate
>300°C
MFCD00007544
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium;octane-1-sulfonate
23669624
≥99%
Crystalline
5324-84-5
C8H17NaO3S
100 g
Soluble in water producing a clear and colorless solution.
[Na+].CCCCCCCCS([O-])(=O)=O
216.27
216.28 (Anhydrous)
HPLC

Chemical Identifiers

5324-84-5
216.27
HRQDCDQDOPSGBR-UHFFFAOYSA-M
23669624
[Na+].CCCCCCCCS([O-])(=O)=O
C8H17NaO3S
MFCD00007544
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
sodium;octane-1-sulfonate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-195-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only