missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium 1-propanesulfonate, 99%, Thermo Scientific Chemicals
$71.96 - $832.74
Chemical Identifiers
CAS | 14533-63-2 |
---|---|
Molecular Formula | C3H7NaO3S |
Molecular Weight (g/mol) | 146.136 |
MDL Number | MFCD00062546 |
InChI Key | NPAWNPCNZAPTKA-UHFFFAOYSA-M |
Synonym | sodium 1-propanesulfonate, sodium propanesulphonate, 1-propanesulfonic acid sodium salt, sodium propane-1-sulfonate, sodium propanesulfonate, acmc-209cuo, ipc-alks-3, c3h7o3s.na, potassium propane-1-sulfonate, propanesulfonic acid sodium salt |
PubChem CID | 4319363 |
IUPAC Name | sodium;propane-1-sulfonate |
SMILES | CCCS(=O)(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2221003
|
Thermo Scientific Chemicals
B2221003 |
1 g |
Each for $71.96
|
|
|||||
AAB2221006
|
Thermo Scientific Chemicals
B2221006 |
5 g |
Each for $239.23
|
|
|||||
AAB2221014
|
Thermo Scientific Chemicals
B2221014 |
25 g |
Each for $832.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14533-63-2 | |
146.136 | |
NPAWNPCNZAPTKA-UHFFFAOYSA-M | |
4319363 | |
CCCS(=O)(=O)[O-].[Na+] |
C3H7NaO3S | |
MFCD00062546 | |
sodium 1-propanesulfonate, sodium propanesulphonate, 1-propanesulfonic acid sodium salt, sodium propane-1-sulfonate, sodium propanesulfonate, acmc-209cuo, ipc-alks-3, c3h7o3s.na, potassium propane-1-sulfonate, propanesulfonic acid sodium salt | |
sodium;propane-1-sulfonate |
Specifications
14533-63-2 | |
MFCD00062546 | |
sodium 1-propanesulfonate, sodium propanesulphonate, 1-propanesulfonic acid sodium salt, sodium propane-1-sulfonate, sodium propanesulfonate, acmc-209cuo, ipc-alks-3, c3h7o3s.na, potassium propane-1-sulfonate, propanesulfonic acid sodium salt | |
CCCS(=O)(=O)[O-].[Na+] | |
146.136 | |
146.14 | |
Sodium 1-propanesulfonate |
C3H7NaO3S | |
1 g | |
NPAWNPCNZAPTKA-UHFFFAOYSA-M | |
sodium;propane-1-sulfonate | |
4319363 | |
99% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-564-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only