Learn More
Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate, Thermo Scientific Chemicals
$89.11 - $1188.84
Chemical Identifiers
CAS | 1191-50-0 |
---|---|
Molecular Formula | C14H29NaO4S |
Molecular Weight (g/mol) | 316.43 |
MDL Number | MFCD00007468 |
InChI Key | UPUIQOIQVMNQAP-UHFFFAOYSA-M |
Synonym | sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate |
PubChem CID | 23665770 |
IUPAC Name | sodium;tetradecyl sulfate |
SMILES | [Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2194103
|
Thermo Scientific Chemicals
B2194103 |
1 g |
Each for $89.11
|
|
||||||
AAB2194106
|
Thermo Scientific Chemicals
B2194106 |
5 g |
Each for $292.19
|
|
||||||
AAB2194114
|
Thermo Scientific Chemicals
B2194114 |
25 g |
Each for $1,188.84
|
|
||||||
Description
Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
1191-50-0 | |
316.43 | |
UPUIQOIQVMNQAP-UHFFFAOYSA-M | |
23665770 | |
[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O |
C14H29NaO4S | |
MFCD00007468 | |
sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate | |
sodium;tetradecyl sulfate |
Specifications
1191-50-0 | |
C14H29NaO4S | |
1 g | |
Hygroscopic | |
Soluble in water. | |
[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O | |
316.43 | |
316.44 | |
Sodium 1-tetradecyl sulfate |
182°C to 192°C | |
MFCD00007468 | |
3578122 | |
sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate | |
UPUIQOIQVMNQAP-UHFFFAOYSA-M | |
sodium;tetradecyl sulfate | |
23665770 | |
95% | |
cont. up to ≈5% sodium methyl sulfate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-737-2
RTECSNumber : XB8660000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only