Learn More
Sodium 2-chloroethanesulfonate hydrate, 98+% (dry wt.), water <10%, Thermo Scientific Chemicals
$65.58 - $737.78
Chemical Identifiers
CAS | 15484-44-3 |
---|---|
Molecular Formula | C2H4ClNaO3S |
Molecular Weight (g/mol) | 166.55 |
MDL Number | MFCD00149546 MFCD00007531 |
InChI Key | BVIXLMYIFZGRBH-UHFFFAOYSA-M |
Synonym | sodium 2-chloroethylsulfonate, 2-chloroethane sulfonic acid sodium salt, sodium .beta.-chloroethanesulfonate, sodium 2-chloroethane-1-sulfonic acid, sodium 2-chloroethanesulfonic acid |
PubChem CID | 54611855 |
SMILES | [Na+].[O-]S(=O)(=O)CCCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1999506
|
Thermo Scientific Chemicals
A1999506 |
5 g |
N/A
|
|
||||||
AAA1999514
|
Thermo Scientific Chemicals
A1999514 |
25 g |
N/A
|
|
||||||
AAA1999522
|
Thermo Scientific Chemicals
A1999522 |
100 g |
N/A
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15484-44-3 | |
166.55 | |
BVIXLMYIFZGRBH-UHFFFAOYSA-M | |
54611855 |
C2H4ClNaO3S | |
MFCD00149546 MFCD00007531 | |
sodium 2-chloroethylsulfonate, 2-chloroethane sulfonic acid sodium salt, sodium .beta.-chloroethanesulfonate, sodium 2-chloroethane-1-sulfonic acid, sodium 2-chloroethanesulfonic acid | |
[Na+].[O-]S(=O)(=O)CCCl |
Specifications
15484-44-3 | |
C2H4ClNaO3S | |
5 g | |
Hygroscopic | |
BVIXLMYIFZGRBH-UHFFFAOYSA-M | |
sodium;2-chloroethanesulfonic acid | |
54611855 | |
≥98% | |
(dry wt.), water <10% |
∼292°C (decomposition) | |
MFCD00149546 MFCD00007531 | |
5649202 | |
sodium 2-chloroethylsulfonate, 2-chloroethane sulfonic acid sodium salt, sodium .beta.-chloroethanesulfonate, sodium 2-chloroethane-1-sulfonic acid, sodium 2-chloroethanesulfonic acid | |
[Na+].[O-]S(=O)(=O)CCCl | |
166.55 | |
166.56 (Anhydrous) | |
Sodium 2-chloroethanesulfonate hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 239-508-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only