missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium, 30wt% dispersion in toluene, <0.1 mm particle size, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 243262500

Catalog No. AC243262500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium
0.9000g/mL
MFCD00085307
4°C
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
[Na]
22.99
CHEBI:26708
30% Min. (Na)
7440-23-5
Na
250 mL
15, 8708
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
5360545
22.98
Dispersion

Chemical Identifiers

7440-23-5
22.99
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
sodium
Na
MFCD00085307
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
CHEBI:26708
[Na]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage. Causes severe skin burns and eye damage. Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure if inhaled. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Store in a dry place. Store in a closed container.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only