missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 4-Acetylbenzenesulfonate 98.0+%, TCI America™

Supplier:  TCI America A14305G

Catalog No. A14305G


Explore available promotions

Description

Description

Specifications

Specifications

Sodium 4-Acetylbenzenesulfonate
White-Yellow
MFCD00007510
4′C-Acetophenonesulfonic Acid Sodium Salt, 4-Acetylbenzenesulfonic Acid Sodium Salt
CC(=O)C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
222.19
222.19
Crystalline Powder
61827-67-6
C8H7NaO4S
5 g
PUWCULQGLSPCMQ-UHFFFAOYSA-M
sodium;4-acetylbenzenesulfonate
23668814
≥98.0% (HPLC,T)

Chemical Identifiers

61827-67-6
222.19
PUWCULQGLSPCMQ-UHFFFAOYSA-M
23668814
CC(=O)C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
C8H7NaO4S
MFCD00007510
4′C-Acetophenonesulfonic Acid Sodium Salt, 4-Acetylbenzenesulfonic Acid Sodium Salt
sodium;4-acetylbenzenesulfonate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions