missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium 4-Amino-1-naphthalenesulfonate Tetrahydrate 95.0+%, TCI America™
Supplier: TCI America A035325G
Specifications
Sodium 4-Amino-1-naphthalenesulfonate Tetrahydrate | |
C10H8NNaO3S | |
25 g | |
JWSRMCCRAJUMLX-UHFFFAOYSA-M | |
sodium;4-aminonaphthalene-1-sulfonate | |
23665720 | |
≥95.0% (N) |
130-13-2 | |
MFCD00044844 | |
Naphthionic Acid Sodium Salt, 1-Naphthylamine-4-sulfonic Acid Sodium Salt, 4-Amino-1-naphthalenesulfonic Acid Sodium Salt, Sodium Naphthionate, Sodium 1-Naphthylamine-4-sulfonate | |
C1=CC=C2C(=C1)C(=CC=C2S(=O)(=O)[O-])N.[Na+] | |
245.228 | |
245.23 | |
Crystalline Powder |
Chemical Identifiers
130-13-2 | |
245.228 | |
JWSRMCCRAJUMLX-UHFFFAOYSA-M | |
23665720 | |
C1=CC=C2C(=C1)C(=CC=C2S(=O)(=O)[O-])N.[Na+] |
C10H8NNaO3S | |
MFCD00044844 | |
Naphthionic Acid Sodium Salt, 1-Naphthylamine-4-sulfonic Acid Sodium Salt, 4-Amino-1-naphthalenesulfonic Acid Sodium Salt, Sodium Naphthionate, Sodium 1-Naphthylamine-4-sulfonate | |
sodium;4-aminonaphthalene-1-sulfonate |
Safety and Handling
EINECSNumber : (4)-0492
TSCA : Yes