missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium 4-aminohippurate hydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1577514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium 4-aminohippurate hydrate | |
123°C to 127°C | |
MFCD00150723 | |
aminohippurate sodium | |
C1=CC(=CC=C1C(=O)NCC(=O)O)N.[Na+] | |
217.18 | |
216.17 (Anhydrous) |
206658-83-5 | |
C9H10N2NaO3+ | |
25 g | |
UNZMYCAEMNVPHX-UHFFFAOYSA-N | |
sodium;2-[(4-aminobenzoyl)amino]acetic acid | |
57465078 | |
98% |
Chemical Identifiers
206658-83-5 | |
217.18 | |
UNZMYCAEMNVPHX-UHFFFAOYSA-N | |
57465078 | |
C1=CC(=CC=C1C(=O)NCC(=O)O)N.[Na+] |
C9H10N2NaO3+ | |
MFCD00150723 | |
aminohippurate sodium | |
sodium;2-[(4-aminobenzoyl)amino]acetic acid |
Safety and Handling
EINECSNumber : 202-309-8
RTECSNumber : MR8157200
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only