missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 4-aminohippurate hydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1577522

 View more versions of this product

Catalog No. AAA1577522


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium 4-aminohippurate hydrate
123°C to 127°C
MFCD00150723
aminohippurate sodium
C1=CC(=CC=C1C(=O)NCC(=O)O)N.[Na+]
217.18
216.17 (Anhydrous)
206658-83-5
C9H10N2NaO3+
100 g
UNZMYCAEMNVPHX-UHFFFAOYSA-N
sodium;2-[(4-aminobenzoyl)amino]acetic acid
57465078
98%

Chemical Identifiers

206658-83-5
217.18
UNZMYCAEMNVPHX-UHFFFAOYSA-N
57465078
C1=CC(=CC=C1C(=O)NCC(=O)O)N.[Na+]
C9H10N2NaO3+
MFCD00150723
aminohippurate sodium
sodium;2-[(4-aminobenzoyl)amino]acetic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 202-309-8

RTECSNumber : MR8157200

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only