missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 99.8%, oiled sticks, wrapped in aluminum foil, Thermo Scientific Chemicals

$81.87 - $141.91

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC390121000
SDS View Documents
Thermo Scientific Chemicals
390121000
100 g
Each for $81.87
 
AC390122500
SDS View Documents
Thermo Scientific Chemicals
390122500
250 g
Each for $141.91
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-23-5
22.99
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
sodium
Na
MFCD00085307
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
CHEBI:26708
[Na]

Specifications

Gray
7440-23-5,8042-47-5
99.8%
Na
100 g
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
[Na]
22.99
CHEBI:26708
99.8%
Sodium
98.0°C
883.0°C
Na
MFCD00085307
15, 8708
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
5360545
23.99
Stick
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only