missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Acetate Anhydrous (Fused Crystals/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical FLS2102

 View more versions of this product

Catalog No. S210-2



Explore available promotions

Description

Description

Specifications

Specifications

Sodium Acetate Anhydrous
127-09-3
7.5 to 9.2
2 kg
C2H3NaO2
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate
517045
82.03
Certified ACS
1.0% max. (at 20°C)
324°C
White
Powder/Solid
≥99 %
CH3COONa
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
[Na+].CC([O-])=O
82.03
CHEBI:32954
≥99%
Pass Test
Glass Bottle

Chemical Identifiers

127-09-3
82.03
VMHLLURERBWHNL-UHFFFAOYSA-M
517045
sodium acetate
C2H3NaO2
MFCD00012459
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
CHEBI:32954
[Na+].CC([O-])=O
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:1

SDS Documents

Documents

Promotions

Promotions