missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Acetate Anhydrous (White Crystals or Granular Powder), Fisher BioReagents
Supplier: Fisher BioReagents FLBP3331
Description
Sodium Acetate is added to DNA reactions to facilitate ethanol precipitation of the DNA.Specifications
Sodium Acetate Anhydrous | |
127-09-3 | |
7.5 to 9.2 | |
1 kg | |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
≥99.0% | |
Glass Bottle |
324°C | |
White | |
Powder/Solid | |
≥99.0 % | |
CH3COONa | |
15, 8709 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium;acetate | |
517045 | |
82.03 | |
1% max. (at 105°C) |
Chemical Identifiers
127-09-3 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
[Na+].CC([O-])=O |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:1
Recommended Storage : RT